missing translation for 'onlineSavingsMsg'
Learn More

EAAT1/GLAST-1/SLC1A3 Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 18 publications

213.00 € - 509.00 €

Specifications

Antigen EAAT1/GLAST-1/SLC1A3
Concentration 1.0 mg/mL
Dilution Western Blot 2 μg/mL, Flow Cytometry 1:200-1:500, ELISA 1:100 - 1:2000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Paraffin 1:50-1:500, Immunohistochemistry-Frozen 1:10-1:500, Flow (Intracellular) 1:500
Applications Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18719563
View Documents
Novus Biologicals
NB100-1869SS
0.025 mL
213.00 €
25µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18796573
View Documents
Novus Biologicals
NB100-1869
0.1 mL
509.00 €
0.10mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

EAAT1/GLAST-1/SLC1A3 Polyclonal specifically detects EAAT1/GLAST-1/SLC1A3 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Flow (Intracellular).
Specifications

Specifications

EAAT1/GLAST-1/SLC1A3
Western Blot 2 μg/mL, Flow Cytometry 1:200-1:500, ELISA 1:100 - 1:2000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Paraffin 1:50-1:500, Immunohistochemistry-Frozen 1:10-1:500, Flow (Intracellular) 1:500
Polyclonal
Rabbit
Cellular Markers, Cognition and Behavior
PBS with 0.02% Sodium Azide
EA6FLJ25094, EAAT1GLAST-1, excitatory amino acid transporter 1, GLAST, GLASTGLAST1, Sodium-dependent glutamate/aspartate transporter 1, solute carrier family 1 (glial high affinity glutamate transporter), member 3, Solute carrier family 1 member 3
SLC1A3
IgG
Affinity Purified
Human, rat and mouse. Based upon 100% immunogen sequence similarity, this antibody is predicted to cross-react with Bovine also.
1.0 mg/mL
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Unconjugated
RUO
Human, Mouse, Rat
P24942
6507
A synthetic peptide made to a C-terminal portion of the rat SLC1A3 protein (between residues 500-542) [Uniprot: P24942]
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
60 kDa
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.