missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EBI2/GPR183 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-27352SS
This item is not returnable.
View return policy
Description
EBI2/GPR183 Polyclonal specifically detects EBI2/GPR183 in Human, Monkey, Primate samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| EBI2/GPR183 | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.05% Sodium Azide | |
| EBI2Epstein-Barr virus induced gene 2 (lymphocyte-specific G protein-coupledreceptor), EBV-induced G-protein coupled receptor 2, Epstein-Barr virus-induced G-protein coupled receptor 2, G protein-coupled receptor 183, G-protein coupled receptor 183 | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| The amino acid sequence used as immunogen is 100% homologous in human, chimpanzee and monkey, and 93% homologous in mouse and rat. | |
| Store at -20C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunohistochemistry (Paraffin) | |
| 1.0 mg/ml | |
| Western Blot 2-4ug/ml∼, Immunohistochemistry-Paraffin 5ug/ml | |
| P32249 | |
| GPR183 | |
| A portion of amino acids 300-350 of human EBI2 was used as the immunogen. | |
| 0.025 mg | |
| GPCR, Immune System Diseases, Immunology | |
| 1880 | |
| Human, Monkey, Primate | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction