missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EBI2/GPR183 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
230.00 € - 500.00 €
Specifications
| Antigen | EBI2/GPR183 |
|---|---|
| Concentration | 1.0 mg/ml |
| Dilution | Western Blot 2-4ug/ml∼, Immunohistochemistry-Paraffin 5ug/ml |
| Applications | Western Blot, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18171472
|
Novus Biologicals
NBP2-27352SS |
0.025 mg |
230.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18796923
|
Novus Biologicals
NBP2-27352 |
500.00 €
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
Description
EBI2/GPR183 Polyclonal specifically detects EBI2/GPR183 in Human, Monkey, Primate samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| EBI2/GPR183 | |
| Western Blot 2-4ug/ml∼, Immunohistochemistry-Paraffin 5ug/ml | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Monkey, Primate | |
| P32249 | |
| 1880 | |
| A portion of amino acids 300-350 of human EBI2 was used as the immunogen. | |
| Primary | |
| Store at -20C. Avoid freeze-thaw cycles. |
| 1.0 mg/ml | |
| Western Blot, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| GPCR, Immune System Diseases, Immunology | |
| PBS with 0.05% Sodium Azide | |
| EBI2Epstein-Barr virus induced gene 2 (lymphocyte-specific G protein-coupledreceptor), EBV-induced G-protein coupled receptor 2, Epstein-Barr virus-induced G-protein coupled receptor 2, G protein-coupled receptor 183, G-protein coupled receptor 183 | |
| GPR183 | |
| IgG | |
| Protein A purified | |
| The amino acid sequence used as immunogen is 100% homologous in human, chimpanzee and monkey, and 93% homologous in mouse and rat. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title