missing translation for 'onlineSavingsMsg'
Learn More

EBOV GP Antibody, Novus Biologicals™

Product Code. p-200072336 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
50 μg
Unit Size:
100µL
50µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18672459 100 μg 100µL
18639557 50 μg 50µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18672459 Supplier Novus Biologicals Supplier No. NBP312747100ul

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

EBOV GP Polyclonal antibody specifically detects EBOV GP in Ebola virus samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen EBOV GP
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:500-1:2000
Formulation 0.2 um filtered solution in PBS
Gene Alias second secreted glycoprotein, small secreted glycoprotein, Spike glycoprotein
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant EBOV GP (Accession#: AAC54887.1; Met1-Gln650)
Purification Method Antigen and protein A Affinity-purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3160774
Target Species Ebola virus
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.