missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EBOV VP24 protein Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-12763-50ul
This item is not returnable.
View return policy
Description
EBOV VP24 protein Polyclonal antibody specifically detects EBOV VP24 protein in Ebola virus samples. It is validated for Western Blot
Specifications
| EBOV VP24 protein | |
| Polyclonal | |
| Western Blot 1:1000-1:5000 | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Ebola virus | |
| Purified |
| Western Blot | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Produced in rabbits immunized with purified, recombinant EBOV VP24 protein (Accession#: YP_003815439.1; Met1-Lys233). | |
| 50 μg | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction