missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EBOV VP24 protein Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
299.00 € - 507.00 €
Specifications
| Antigen | EBOV VP24 protein |
|---|---|
| Dilution | Western Blot 1:1000-1:5000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18660238
|
Novus Biologicals
NBP3-12763-100ul |
100 μg |
507.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18621837
|
Novus Biologicals
NBP3-12763-50ul |
50 μg |
299.00 €
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
EBOV VP24 protein Polyclonal antibody specifically detects EBOV VP24 protein in Ebola virus samples. It is validated for Western BlotSpecifications
| EBOV VP24 protein | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Ebola virus | |
| Produced in rabbits immunized with purified, recombinant EBOV VP24 protein (Accession#: YP_003815439.1; Met1-Lys233). | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot 1:1000-1:5000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| 0.2 um filtered solution in PBS | |
| IgG | |
| Antigen and protein A Affinity-purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title