missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EGFR Mouse, Clone: 31G7, enQuireBio™
Mouse Monoclonal Antibody
199.00 € - 485.00 €
Specifications
Antigen | EGFR |
---|---|
Clone | 31G7 |
Concentration | 200μg/mL |
Applications | Flow Cytometry, Immunofluorescence, Immunohistochemistry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15929927
|
EnquireBio
1956-MSM4-P1 |
100 μg |
485.00 €
100µg |
Estimated Shipment: 04-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15919927
|
EnquireBio
1956-MSM4-P0 |
20 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
EGFR | |
200μg/mL | |
Monoclonal | |
Mouse | |
10mM PBS with 0.05% BSA and 0.05% azide | |
1956 | |
Human EGFR purified from A431 cells | |
Primary | |
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. | |
∽170kDa (wild type), ∽145kDa (vIII variant) |
31G7 | |
Flow Cytometry, Immunofluorescence, Immunohistochemistry | |
Unconjugated | |
RUO | |
NP_001333828 | |
EGFR | |
IgG1 κ | |
Protein A/G Purified | |
Cell surface | |
A431 cells. Placenta, Breast, Colon or Bladder cancer |