missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EGR3 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
190.00 € - 463.00 €
Specifications
| Antigen | EGR3 |
|---|---|
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18626332
|
Novus Biologicals
NBP2-92512-0.02ml |
0.02 mL |
190.00 €
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18622852
|
Novus Biologicals
NBP2-92512-0.1ml |
0.1 mL |
463.00 €
0.01mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
EGR3 Polyclonal antibody specifically detects EGR3 in Human, Mouse samples. It is validated for Western BlotSpecifications
| EGR3 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Neuroscience, Signal Transduction | |
| PBS with 50% glycerol, pH7.3. | |
| 1960 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse | |
| early growth response 3, early growth response protein 3, EGR-3, MGC138484, PILOTZinc finger protein pilot | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-250 of human EGR3 (NP_004421.2). MTGKLAEKLPVTMSSLLNQLPDNLYPEEIPSALNLFSGSSDSVVHYNQMATENVMDIGLTNEKPNPELSYSGSFQPAPGNKTVTYLGKFAFDSPSNWCQDNIISLMSAGILGVPPASGALSTQTSTASMVQPPQGDVEAMYPALPPYSNCGDLYSEPVSFHDPQGNPGLAYSPQDYQSAKPALDSNLFPMIPDYNLYHHPNDMGSIPEHKPFQGMDPIRVNPPPITPLETIKAFKDKQIHPGFGSLPQPP | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title