Learn More
eIF4G1, p Ser1231 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 3 publications
Brand: Novus Biologicals NBP1-04965
This item is not returnable.
View return policy
Description
eIF4G1 (p Ser1231) Polyclonal antibody specifically detects eIF4G1 (p Ser1231) in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin).
Specifications
| EIF4G1 | |
| 220 kDa |