missing translation for 'onlineSavingsMsg'
Learn More
Learn More
eIF4G1, p Ser1231 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 3 publications
176.00 € - 493.00 €
Specifications
| Concentration | 1.0 mg/mL |
|---|---|
| Dilution | Western Blot 1:500-1:1000, Immunohistochemistry 1:50-1:100, Immunohistochemistry-Paraffin 1:50-1:100 |
| Applications | Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18227622
|
Novus Biologicals
NBP1-04965-0.025ML |
0.025 mL |
176.00 €
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18736193
|
Novus Biologicals
NBP1-04965 |
493.00 €
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
Description
eIF4G1 Polyclonal specifically detects eIF4G1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| 1.0 mg/mL | |
| Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| Human | |
| DKFZp686A1451, EIF4F, eIF-4G 1, EIF-4G1, EIF4-gamma, eIF-4-gamma 1, EIF4GI, EIF4Gp220, eucaryotic translation initiation factor 4G, eukaryotic translation initiation factor 4 gamma 1, eukaryotic translation initiation factor 4 gamma, 1, P220 | |
| EIF4G1 | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot 1:500-1:1000, Immunohistochemistry 1:50-1:100, Immunohistochemistry-Paraffin 1:50-1:100 | |
| Polyclonal | |
| Rabbit | |
| Cancer, DNA Repair, DNA replication Transcription Translation and Splicing, mTOR Pathway, Phospho Specific | |
| Q04637 | |
| 1981 | |
| IgG | |
| Affinity Purified | |
| 220 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title