missing translation for 'onlineSavingsMsg'
Learn More

ELN484228, TRC

Product Code. 30380470
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30380470 1 g
30385714 500 mg
30388235 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30380470 Supplier Toronto Research Chemicals Supplier No. TRCE5000051G
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 312-63-0
Molecular Formula C12 H10 F N O2 S
Molecular Weight (g/mol) 251.277
Synonym ELN484228, 4'-Fluorobenzenesulfonanilide, N-(p-Fluorophenyl)benzenesulfonamide, NSC 164389
IUPAC Name N-(4-fluorophenyl)benzenesulfonamide
SMILES Fc1ccc(NS(=O)(=O)c2ccccc2)cc1

Specifications

Quantity 1 g
InChI Formula InChI=1S/C12H10FNO2S/c13-10-6-8-11(9-7-10)14-17(15,16)12-4-2-1-3-5-12/h1-9,14H
Formula Weight 251.042
Chemical Name or Material N-(4-Fluorophenyl)benzenesulfonamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.