missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
ELOVL5 Monoclonal antibody specifically detects ELOVL5 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
Specifications
Specifications
| Antigen | ELOVL5 |
| Applications | Western Blot, Immunofluorescence |
| Classification | Monoclonal |
| Clone | S07-7B3 |
| Concentration | 0.3 mg/mL |
| Conjugate | Unconjugated |
| Formulation | 50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA |
| Gene Alias | dJ483K16.1, EC 2.3.1.n8, elongation of very long chain fatty acids protein 5, ELOVL family member 5, elongation of long chain fatty acids (FEN1/Elo2, ELOVL2,3-keto acyl-CoA synthase ELOVL5, Fatty acid elongase 1, hELO1, HELO1homolog of yeast long chain polyunsaturated fatty acid elongation enzyme 2, homolog of yeast long chain polyunsaturated fatty acid elongatio, SUR4/Elo3-like, yeast) |
| Host Species | Rabbit |
| Immunogen | A synthetic peptide of human ELOVL5 (Uniprot # Q9NYP7) |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?