missing translation for 'onlineSavingsMsg'
Learn More

Emtricitabine Sulfoxide, Mikromol™

Product Code. 17912386
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17912386 25 mg 25mg
17971796 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17912386 Supplier Mikromol Supplier No. MM1325.020025
 Restricted Item
Estimated Shipment: 04-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Emtricitabine Sulfoxide
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Emtricitabine S-Oxide (Mixture of Diastereomers)
CAS 152128-77-3
Analyte or Component Names Emtricitabine S-Oxide (Mixture of Diastereomers)
Shipping Condition Room Temperature
Molecular Formula C8 H10 F N3 O4 S
Certifications/Compliance ISO 9001
Synonym 2(1H)-Pyrimidinone, 4-amino-5-fluoro-1-[(2R,5S)-2-(hydroxymethyl)-3-oxido-1,3-oxathiolan-5-yl]-, 1,3-Oxathiolane, 2(1H)-pyrimidinone deriv., 4-Amino-5-fluoro-1-[(2R,5S)-2-(hydroxymethyl)-3-oxido-1,3-oxathiolan-5-yl]-2(1H)-pyrimidinone, 2(1H)-Pyrimidinone, 4-amino-5-fluoro-1-[2-(hydroxymethyl)-1,3-oxathiolan-5-yl]-, S-oxide, [2R-(2alpha,5alpha)]-, 2(1H)-Pyrimidinone, 4-amino-5-fluoro-1-[2-(hydroxymethyl)-3-oxido-1,3-oxathiolan-5-yl]-, [2R-(2alpha,5alpha)]- (9CI)
InChI Formula InChI=1S/C8H10FN3O4S/c9-4-1-12(8(14)11-7(4)10)5-3-17(15)6(2-13)16-5/h1,5-6,13H,2-3H2,(H2,10,11,14)/t5-,6+,17?/m0/s1
SMILES NC1=NC(=O)N(C=C1F)[C@@H]2CS(=O)[C@H](CO)O2
IUPAC Name 4-amino-5-fluoro-1-[(2R,5S)-2-(hydroxymethyl)-3-oxo-1,3-oxathiolan-5-yl]pyrimidin-2-one
Molecular Weight (g/mol) 263.25
Quantity 25 mg
Formula Weight 263.0376 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.