missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
EN1/Engrailed 1 Monoclonal antibody specifically detects EN1/Engrailed 1 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunoprecipitation
Specifications
Specifications
| Antigen | EN1/Engrailed 1 |
| Applications | Western Blot, Flow Cytometry, Immunoprecipitation |
| Classification | Monoclonal |
| Clone | SR1987 |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:500-1:2000, Flow Cytometry 1:100, Immunoprecipitation 1:50 |
| Formulation | PBS (pH 7.4), 150 mM NaCl, 50% Glycerol |
| Gene Alias | engrailed homeobox 1, engrailed homolog 1, Homeobox protein en-1, homeobox protein engrailed-1, hu-En-1 |
| Host Species | Rabbit |
| Immunogen | A synthesized peptide derived from human EN1/Engrailed 1 (Uniprot #: Q05925) |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?