missing translation for 'onlineSavingsMsg'
Learn More

Endoglin/CD105 Antibody (ENG/1327), PerCP, R&D Systems™

Product Code. 18620688 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18620688 100 μg 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18620688 Supplier Novus Biologicals Supplier No. NBP254517PCP
  • ONLINE EXCLUSIVE PRICE
    666.04 €
    572.25 € / 100µL
    Save 93.79 €
    14% Off
 Restricted Item
Estimated Shipment: 01-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Novus product, part of R&D Systems. Mouse Monoclonal Antibody

Endoglin/CD105 Monoclonal antibody specifically detects Endoglin/CD105 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/ Immunofluorescence, CyTOF
TRUSTED_SUSTAINABILITY

Specifications

Antigen Endoglin/CD105
Applications Western Blot, Flow Cytometry, Immunofluorescence, CyTOF
Classification Monoclonal
Clone ENG/1327
Conjugate PerCP
Formulation PBS
Gene Alias CD105, CD105 antigen, endoglin, ENDOsler-Rendu-Weber syndrome 1, HHT1FLJ41744, ORW, ORW1
Host Species Mouse
Immunogen Recombinant human Endoglin/CD105 protein fragment (aa74-251) (exact sequence is proprietary) (Uniprot: P17813)
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cancer, Cell Biology, Cellular Markers, Endothelial Cell Markers, Extracellular Matrix, Hypoxia, Immunology, Mesenchymal Stem Cell Markers, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 2022
Target Species Human
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG1 κ
Product Line Novus
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.