missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Endorepellin/Perlecan/Heparan Sulfate Proteoglycan Antibody (A76) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-05170-0.025MG
This item is not returnable.
View return policy
Description
Endorepellin/Perlecan/Heparan Sulfate Proteoglycan Monoclonal specifically detects Endorepellin/Perlecan/Heparan Sulfate Proteoglycan in Human, Bovine samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunoaffinity Purification.
Specifications
| Endorepellin/Perlecan/Heparan Sulfate Proteoglycan | |
| Monoclonal | |
| 1.0 mg/mL | |
| Western Blot 1:100-1:2000, ELISA 1:8000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500 | |
| P98160 | |
| HSPG2 | |
| Lysed bovine corneal endothelial cells and extracellular matrix | |
| 0.025 mg | |
| Primary | |
| Highly specific for perlecan. There is no evidence for cross-reactivity with other connective tissue proteins (vitronectin, fibronectin, elastin, collagen, laminin). | |
| Store at 4C in the dark. | |
| IgG1 |
| Immunoprecipitation, Immunohistochemistry (Frozen), Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry, ELISA, Immunofluorescence | |
| A76 | |
| Unconjugated | |
| 10mM Phosphate (pH 7.4) and 0.5M NaCl with 0.05% Sodium Azide | |
| basement membrane-specific heparan sulfate proteoglycan core protein, endorepellin (domain V region), heparan sulfate proteoglycan 2, HSPG, perlecan, perlecan proteoglycan, PLCSchwartz-Jampel syndrome 1 (chondrodystrophic myotonia), PRCAN, SJA, SJS, SJS1 | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| 3339 | |
| Human, Bovine | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction