missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Endorepellin/Perlecan/Heparan Sulfate Proteoglycan Antibody (A76) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
232.00 € - 592.00 €
Specifications
| Antigen | Endorepellin/Perlecan/Heparan Sulfate Proteoglycan |
|---|---|
| Clone | A76 |
| Concentration | 1.0 mg/mL |
| Dilution | Western Blot 1:100-1:2000, ELISA 1:8000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500 |
| Applications | Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18243350
|
Novus Biologicals
NBP1-05170 |
100 μg |
592.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18054516
|
Novus Biologicals
NBP1-05170-0.025MG |
0.025 mg |
232.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Endorepellin/Perlecan/Heparan Sulfate Proteoglycan Monoclonal specifically detects Endorepellin/Perlecan/Heparan Sulfate Proteoglycan in Human, Bovine samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunoaffinity Purification.Specifications
| Endorepellin/Perlecan/Heparan Sulfate Proteoglycan | |
| 1.0 mg/mL | |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
| Unconjugated | |
| Mouse | |
| Human, Bovine | |
| basement membrane-specific heparan sulfate proteoglycan core protein, endorepellin (domain V region), heparan sulfate proteoglycan 2, HSPG, perlecan, perlecan proteoglycan, PLCSchwartz-Jampel syndrome 1 (chondrodystrophic myotonia), PRCAN, SJA, SJS, SJS1 | |
| HSPG2 | |
| IgG1 | |
| Protein A or G purified | |
| Highly specific for perlecan. There is no evidence for cross-reactivity with other connective tissue proteins (vitronectin, fibronectin, elastin, collagen, laminin). |
| A76 | |
| Western Blot 1:100-1:2000, ELISA 1:8000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500 | |
| Monoclonal | |
| Purified | |
| RUO | |
| P98160 | |
| 3339 | |
| Lysed bovine corneal endothelial cells and extracellular matrix | |
| Primary | |
| Store at 4C in the dark. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title