missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Eosinophil Chemotactic Factor Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5122277
This item is not returnable.
View return policy
Description
CCL11 (C-C Motif Chemokine Ligand 11) is a Protein Coding gene. Diseases associated with CCL11 include Asthma and Human Immunodeficiency Virus Type 1. Among its related pathways are ERK Signaling and Immune response_Oncostatin M signaling via JAK-Stat in human cells. Gene Ontology (GO) annotations related to this gene include chemokine activity. An important paralog of this gene is CCL2. This antimicrobial gene is one of several chemokine genes clustered on the q-arm of chromosome 17. Chemokines form a superfamily of secreted proteins involved in immunoregulatory and inflammatory processes. The superfamily is divided into four subfamilies based on the arrangement of the N-terminal cysteine residues of the mature peptide. This chemokine, a member of the CC subfamily, displays chemotactic activity for eosinophils, but not mononuclear cells or neutrophils. This eosinophil-specific chemokine is thought to be involved in eosinophilic inflammatory diseases such as atopic dermatitis, allergic rhinitis, asthma and parasitic infections.
Specifications
| Eosinophil Chemotactic Factor | |
| Polyclonal | |
| Unconjugated | |
| CCL11 | |
| ECF | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 6356 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry, Western Blot, Immunocytochemistry | |
| 0.5 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| P51671 | |
| CCL11 | |
| Gly24-Pro97 (Accession Q6I9T4), with N-terminal His Tag. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction