missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Eosinophil Chemotactic Factor Polyclonal Antibody, Invitrogen™
GREENER_CHOICE

Rabbit Polyclonal Antibody

134.00 € - 453.00 €

Specifications

Antigen Eosinophil Chemotactic Factor
Concentration 0.5 mg/mL
Applications Immunohistochemistry, Western Blot, Immunocytochemistry
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
17763674
View Documents
Invitrogen™
PA5122278
20 μL
134.00 €
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
17753674
GREENER_CHOICE
View Documents
Invitrogen™
PA5122277
100 μL
453.00 €
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

CCL11 (C-C Motif Chemokine Ligand 11) is a Protein Coding gene. Diseases associated with CCL11 include Asthma and Human Immunodeficiency Virus Type 1. Among its related pathways are ERK Signaling and Immune response_Oncostatin M signaling via JAK-Stat in human cells. Gene Ontology (GO) annotations related to this gene include chemokine activity. An important paralog of this gene is CCL2. This antimicrobial gene is one of several chemokine genes clustered on the q-arm of chromosome 17. Chemokines form a superfamily of secreted proteins involved in immunoregulatory and inflammatory processes. The superfamily is divided into four subfamilies based on the arrangement of the N-terminal cysteine residues of the mature peptide. This chemokine, a member of the CC subfamily, displays chemotactic activity for eosinophils, but not mononuclear cells or neutrophils. This eosinophil-specific chemokine is thought to be involved in eosinophilic inflammatory diseases such as atopic dermatitis, allergic rhinitis, asthma and parasitic infections.
Specifications

Specifications

Eosinophil Chemotactic Factor
Immunohistochemistry, Western Blot, Immunocytochemistry
Unconjugated
Rabbit
Human
P51671
6356
Gly24-Pro97 (Accession Q6I9T4), with N-terminal His Tag.
Primary
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
CCL11
0.5 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
ECF
CCL11
IgG
Antigen affinity chromatography, Protein A
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.