Learn More
Invitrogen™ Epsin 1 Recombinant Rabbit Monoclonal Antibody (23GB1525), Invitrogen™
GREENER_CHOICE
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA553116
 View more versions of this product
Product Code. 30087935
  • 446.00 € / 100µL
Description
This gene encodes a member of the epsin protein family. The encoded protein binds clathrin and is involved in the endocytosis of clathrin-coated vesicles. Loss of function of this gene is associated with reduced tumor growth and progression in certain cancer types.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Epsin 1
Recombinant Monoclonal
0.4 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Q9Y6I3
EPN1
A synthesized peptide derived from human Epsin 1 (450-490AA).
100 μL
Primary
Human
Antibody
IgG
Flow Cytometry, Western Blot, Immunocytochemistry
23GB1525
Unconjugated
EPN1
AA269831; EH domain-binding mitotic phosphoprotein; Epn1; EPS-15-interacting protein 1; Epsin 1; epsin-1; hypothetical protein LOC508199; Ibp1; Intersectin-EH-binding protein 1; LOW QUALITY PROTEIN: epsin-1
Rabbit
Affinity chromatography
RUO
29924
-20°C