missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Epsin 1 Recombinant Rabbit Monoclonal Antibody (23GB1525), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
186.00 € - 446.00 €
Specifications
| Antigen | Epsin 1 |
|---|---|
| Clone | 23GB1525 |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30087935
![]() |
Invitrogen™
MA553116 |
100 μL |
446.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30088245
|
Invitrogen™
MA553117 |
20 μL |
186.00 €
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Epsin 1 Recombinant Monoclonal antibody specifically detects Epsin 1 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Western BlotSpecifications
| Epsin 1 | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human | |
| Q9Y6I3 | |
| 29924 | |
| IgG | |
| Affinity chromatography | |
| Antibody |
| 23GB1525 | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| AA269831; EH domain-binding mitotic phosphoprotein; Epn1; EPS-15-interacting protein 1; Epsin 1; epsin-1; hypothetical protein LOC508199; Ibp1; Intersectin-EH-binding protein 1; LOW QUALITY PROTEIN: epsin-1 | |
| A synthesized peptide derived from human Epsin 1 (450-490AA). | |
| Primary | |
| -20°C | |
| EPN1 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title