missing translation for 'onlineSavingsMsg'
Learn More

EQTN Antibody - BSA Free, Novus Biologicals™

Product Code. 30558657 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30558657 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30558657 Supplier Novus Biologicals Supplier No. NBP347738

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

EQTN Polyclonal antibody specifically detects EQTN in Human samples. It is validated for ELISA,Western Blot,Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen EQTN
Applications ELISA, Western Blot, Immunocytochemistry/Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:500-1:2000, ELISA, Immunocytochemistry/ Immunofluorescence 1:10-1:100
Formulation PBS, 50% Glycerol, pH (7.3)
Gene Alias acrosome formation associated factor, acrosome formation-associated factor, AFAF, C9orf11, chromosome 9 open reading frame 11, equatorin, equatorin, sperm acrosome associated, SPACA8
Host Species Rabbit
Immunogen chromosome 9 open reading frame 11. (Uniprot# Q9NQ60)
Purification Method Immunogen affinity purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54586
Target Species Human
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.