missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ERK1/ERK2, p Tyr204 Antibody (SR1718), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-21522-50ul
This item is not returnable.
View return policy
Description
ERK1/ERK2 Monoclonal antibody specifically detects ERK1/ERK2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunoprecipitation
Specifications
| ERK1/ERK2 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| A synthesized peptide derived from human phospho-ERK1 [Tyr204]/ERK2 [Tyr187] (Uniprot #: P27361, P28482) | |
| 50 μg | |
| Primary | |
| Store at -20 degrees C. Avoid freeze/thaw cycles. | |
| IgG |
| Western Blot, Immunoprecipitation | |
| SR1718 | |
| Western Blot 1:500-1:2000, Immunoprecipitation 1:50 | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction