missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ERK1/ERK2, p Tyr204 Antibody (SR1718), Novus Biologicals™
Rabbit Monoclonal Antibody
338.00 € - 449.00 €
Specifications
| Antigen | ERK1/ERK2 |
|---|---|
| Clone | SR1718 |
| Dilution | Western Blot 1:500-1:2000, Immunoprecipitation 1:50 |
| Applications | Western Blot, Immunoprecipitation |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18690555
|
Novus Biologicals
NBP3-21522-50ul |
50 μg |
338.00 €
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18678854
|
Novus Biologicals
NBP3-21522-100ul |
100 μg |
449.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ERK1/ERK2 Monoclonal antibody specifically detects ERK1/ERK2 in Human, Mouse, Rat samples. It is validated for Western Blot, ImmunoprecipitationSpecifications
| ERK1/ERK2 | |
| Western Blot 1:500-1:2000, Immunoprecipitation 1:50 | |
| Monoclonal | |
| Purified | |
| RUO | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| IgG | |
| Affinity purified |
| SR1718 | |
| Western Blot, Immunoprecipitation | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| A synthesized peptide derived from human phospho-ERK1 [Tyr204]/ERK2 [Tyr187] (Uniprot #: P27361, P28482) | |
| Primary | |
| Store at -20 degrees C. Avoid freeze/thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title