missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
ERV3-1 Polyclonal antibody specifically detects ERV3-1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
Specifications
| Antigen | ERV3-1 |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Western Blot 0.04-0.4 μg/ml, Immunohistochemistry 1:500 - 1:1000, Immunohistochemistry-Paraffin 1:500 - 1:1000 |
| Formulation | PBS (pH 7.2) and 40% Glycerol |
| Gene Alias | Endogenous Retroviral Sequence 3, Endogenous Retroviral Sequence 3 (Includes Zinc Finger Protein H-Plk/HPF9), Endogenous Retrovirus Group 3 Member 1, Endogenous Retrovirus Group 3 Member 1 Env Polyprotein, Endogenous Retrovirus Group 3 Member 1, Envelope, Endogenous Retrovirus Group 3, Member 1, envelope polyprotein, ERV3, ERV3 Envelope Protein, ERV-3 Envelope Protein, ERV3-1 Envelope Protein, ERV-R, ERV-R Envelope Protein, HERVR, HERV-R, HERV-R_7q21.2 Provirus Ancestral Env Polyprotein |
| Host Species | Rabbit |
| Immunogen | This antibody was developed against a recombinant protein corresponding to amino acids: QLAENIASSLHVASCYVCGGMNMGDQWPWEARELMPQDNFTLTASSLEPAPSSQSIWFLKTSIIGKFCIARWGKAFTDPVGELTCLGQQYYNETLGKTLWRGKSNNSESPHPSPFSRFPSLNHSWYQLEA |
| Purification Method | Protein A purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?