missing translation for 'onlineSavingsMsg'
Learn More

Estra-1,3,5(10),9(11)-tetraene-3,17beta-diol (9,11-Didehydroestradiol), Mikromol™

Product Code. 18062060
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18062060 25 mg 25mg
18061960 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18062060 Supplier Mikromol Supplier No. MM0300.020025

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Estra-1,3,5(10),9(11)-tetraene-3,17beta-diol (9,11-Didehydroestradiol)
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 9,11-Didehydroestradiol
CAS 791-69-5
Analyte or Component Names 9,11-Didehydroestradiol
Shipping Condition Ice pack (-20°C)
Molecular Formula C18 H22 O2
Certifications/Compliance ISO 9001
Synonym Estra-1,3,5(10),9(11)-tetraene-3,17beta-diol, Estradiol Hemihydrate Imp. D (EP), 9,11-Didehydroestradiol, Estradiol Hemihydrate Impurity D, Estradiol Impurity D
InChI Formula InChI=1S/C18H22O2/c1-18-9-8-14-13-5-3-12(19)10-11(13)2-4-15(14)16(18)6-7-17(18)20/h3,5,8,10,15-17,19-20H,2,4,6-7,9H2,1H3/t15-,16+,17+,18+/m1/s1
SMILES C[C@]12CC=C3[C@@H](CCc4cc(O)ccc34)[C@@H]1CC[C@@H]2O
IUPAC Name (8S,13S,14S,17S)-13-methyl-6,7,8,12,14,15,16,17-octahydrocyclopenta[a]phenanthrene-3,17-diol
Molecular Weight (g/mol) 270.37
Quantity 25 mg
Formula Weight 270.162 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.