missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Estradiol Cypionate, LoGiCal
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Specifications
Specifications
| Chemical Name or Material | Estradiol Cypionate |
| CAS | 313-06-4 |
| Analyte or Component Names | Estradiol Cypionate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C26 H36 O3 |
| Certifications/Compliance | ISO 9001 |
| Synonym | Estradiol, 17-cyclopentanepropionate (6CI,7CI,8CI), Cyclopentanepropionic acid, 17-ester with estradiol, Depgynogen, Depo-Estradiol, Depo-estradiol cyclopentylpropionate, Depoestradiol, Depoestradiol cypionate, Depofemin, ECP, Estradep, Estradiol 17-cyclopentylpropionate, Estradiol 17-cypionate, Estradiol cyclopentylpropionate, Estradiol cypionate, Estradiolum valerianicum, NSC 3354, Cyclopentanepropionic acid, 3-hydroxyestra-1,3,5(10)-trien-17beta-yl ester (8CI), 17beta-Estradiol 17-cyclopentylpropionate, Estradiol 17beta-cyclopentanepropionate, Estradiol 17beta-cyclopentylpropionate, Estradiol 17beta-cypionate, Estra-1,3,5(10)-triene-3,17-diol (17beta)-, 17-cyclopentanepropanoate |
| InChI Formula | InChI=1S/C26H36O3/c1-26-15-14-21-20-10-8-19(27)16-18(20)7-9-22(21)23(26)11-12-24(26)29-25(28)13-6-17-4-2-3-5-17/h8,10,16-17,21-24,27H,2-7,9,11-15H2,1H3/t21-,22-,23+,24+,26+/m1/s1 |
| Solution Type | Neat |
| SMILES | C[C@]12CC[C@H]3[C@@H](CCc4cc(O)ccc34)[C@@H]1CC[C@@H]2OC(=O)CCC5CCCC5 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction