missing translation for 'onlineSavingsMsg'
Learn More

Estradiol Hemihydrate (17beta-Estradiol Hemihydrate) 1.0 mg/ml in Acetonitrile (anhydrous substance), LoGiCal

Product Code. 30462057
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30462057 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30462057 Supplier LoGiCal Supplier No. LGCAMP0300.0011

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

LoGiCal delivers certified reference materials you can trust as neats, native solutions and stable isotope labelled solutions, with precise documentation and expert support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.

Estradiol Hemihydrate (17beta-Estradiol Hemihydrate) 1.0 mg/ml in Acetonitrile (anhydrous substance)
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Estradiol Hemihydrate
CAS 35380-71-3
Analyte or Component Names Estradiol Hemihydrate
Shipping Condition Room Temperature
Molecular Formula 2 C18 H24 O2 . H2 O
Certifications/Compliance ISO 17034
Synonym Estradiol Benzoate Imp. A (EP), Estradiol Valerate Imp. A (EP), Estriol Imp. D (EP), Ethinylestradiol Imp. D (EP), Estradiol Hemihydrate, Estra-1,3,5(10)-triene-3,17-diol (17 beta)-, hydrate (2:1) (9 CI), Estra-1,3,5(10)-triene-3,17-diol (17 beta)-, hydrate (2:1) (9 CI), Estra-1,3,5(10)-triene-3,17-diol (17 beta)-, hydrate (2:1) (9 CI), Estra-1,3,5(10)-triene-3,17-diol (17 beta)-, hydrate (2:1) (9 CI), Estra-1,3,5(10)-triene-3,17-diol (17 beta)-, hydrate (2:1) (9 CI), Estra-1,3,5(10)-triene-3,17-diol (17 beta)-, hydrate (2:1) (9 CI), Estra-1,3,5(10)-triene-3,17-diol (17 beta)-, hydrate (2:1) (9 CI), Estra-1,3,5(10)-triene-3,17-diol (17 beta)-, hydrate (2:1) (9 CI), Estra-1,3,5(10)-triene-3,17-diol (17 beta)-, hydrate (2:1) (9 CI), Estra-1,3,5(10)-triene-3,17-diol (17 beta)-, hydrate (2:1) (9 CI), Estra-1,3,5(10)-triene-3,17-diol (17 beta)-, hydrate (2:1) (9 CI)
InChI Formula InChI=1 S/2C18H24O2.H2O/c2*1-18-9-8-14-13-5-3-12(19)10-11(13)2-4-15(14)16(18)6-7-17(18)20;/h2*3,5,10,14-17,19-20 H,2,4,6-9H2,1H3;1H2/t2*14-,15-,16+,17+,18+;/m11./s1
SMILES O.C[C@]12 CC[C@H]3 [C@@H](CCc4cc(O)ccc34)[C@@H]1 CC[C@@H]2 O.C[C@]56 CC[C@H]7 [C@@H](CCc8cc(O)ccc78)[C@@H]5 CC[C@@H]6 O
IUPAC Name (8 R,9 S,13 S,14 S,17 S)-13-methyl-6,7,8,9,11,12,14,15,16,17-decahydrocyclopenta[a]phenanthrene-3,17-diol;hydrate
Solvent or Matrix Acetonitrile
Industry Type Forensics
Molecular Weight (g/mol) 562.78
Quantity 1 mL
Formula Weight 562.3658 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.