missing translation for 'onlineSavingsMsg'
Learn More

Ethionamide, Mikromol™

Product Code. 17913556
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17913556 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17913556 Supplier Mikromol Supplier No. MM0845.00

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Ethionamide
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Ethionamide
CAS 536-33-4
Analyte or Component Names Ethionamide
Shipping Condition Room Temperature
Molecular Formula C8 H10 N2 S
Certifications/Compliance ISO 17025
Synonym Ethionamide
InChI Formula InChI=1S/C8H10N2S/c1-2-7-5-6(8(9)11)3-4-10-7/h3-5H,2H2,1H3,(H2,9,11)
SMILES CCc1cc(ccn1)C(=S)N
IUPAC Name 2-ethylpyridine-4-carbothioamide
Molecular Weight (g/mol) 166.24
Quantity 250 mg
Formula Weight 166.0565 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.