missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Parahydroxybenzoate, LoGiCal

Product Code. 30458881
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30458881 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30458881 Supplier LoGiCal Supplier No. LGCFOR0498.00
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

LoGiCal delivers certified reference materials you can trust as neats, native solutions and stable isotope labelled solutions, with precise documentation and expert support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.

Ethyl Parahydroxybenzoate
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Ethyl Parahydroxybenzoate
CAS 120-47-8
Analyte or Component Names Ethyl Parahydroxybenzoate
Shipping Condition Room Temperature
Molecular Formula C9 H10 O3
Certifications/Compliance ISO 9001
Synonym Ethyl Parahydroxybenzoate, Ethyl 4-Hydroxybenzoate, Butyl Parahydroxybenzoate Imp. C (EP), Propyl Parahydroxybenzoate Imp. C (EP), Methyl Parahydroxybenzoate Imp. B (EP), 4-(Ethoxycarbonyl)phenol, 4-Carbethoxyphenol, 4-Hydroxybenzoic acid ethyl ester, Aseptin A, Aseptine A, Aseptoform E, Bonomold OE, Easeptol, Ethyl 4-hydroxybenzoate, Ethyl Butex, Ethyl nipagin, Ethyl p-hydroxybenzoate, Ethyl parasept, Ethylparaben, Mekkings E, Mycocten, NSC 23514, NSC 8510, Napagin A, Nipagin A, Nipazin A, Sobrol A, Solbrol A, Tegosept E, p-(Ethoxycarbonyl)phenol, p-Carbethoxyphenol, p-Hydroxybenzoate ethyl ester, p-Hydroxybenzoic acid ethyl ester, ethyl paraben, 4-Hydroxybenzoic acid-ethyl ester, Butyl Parahydroxybenzoate Impurity C, Methyl Parahydroxybenzoate Impurity B, Propyl Parahydroxybenzoate Impurity C
InChI Formula InChI=1 S/C9H10O3/c1-2-12-9(11)7-3-5-8(10)6-4-7/h3-6,10 H,2H2,1H3
SMILES CCOC(=O)c1ccc(O)cc1
IUPAC Name ethyl 4-hydroxybenzoate
Industry Type Forensics
Molecular Weight (g/mol) 166.17
Quantity 10 mg
Formula Weight 166.063 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.