missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Parahydroxybenzoate, Mikromol™

Product Code. 17957484
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17957484 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17957484 Supplier Mikromol Supplier No. MM0498.000250
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Ethyl Parahydroxybenzoate
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Ethyl Parahydroxybenzoate
CAS 120-47-8
Analyte or Component Names Ethyl Parahydroxybenzoate
Shipping Condition Room Temperature
Molecular Formula C9 H10 O3
Certifications/Compliance ISO 17034
Synonym Ethyl Parahydroxybenzoate, Ethyl 4-Hydroxybenzoate, Butyl Parahydroxybenzoate Imp. C (EP), Propyl Parahydroxybenzoate Imp. C (EP), Methyl Parahydroxybenzoate Imp. B (EP), 4-(Ethoxycarbonyl)phenol, 4-Carbethoxyphenol, 4-Hydroxybenzoic acid ethyl ester, Aseptin A, Aseptine A, Aseptoform E, Bonomold OE, Easeptol, Ethyl 4-hydroxybenzoate, Ethyl Butex, Ethyl nipagin, Ethyl p-hydroxybenzoate, Ethyl parasept, Ethylparaben, Mekkings E, Mycocten, NSC 23514, NSC 8510, Napagin A, Nipagin A, Nipazin A, Sobrol A, Solbrol A, Tegosept E, p-(Ethoxycarbonyl)phenol, p-Carbethoxyphenol, p-Hydroxybenzoate ethyl ester, p-Hydroxybenzoic acid ethyl ester, ethyl paraben, 4-Hydroxybenzoic acid-ethyl ester, Butyl Parahydroxybenzoate Impurity C, Methyl Parahydroxybenzoate Impurity B, Propyl Parahydroxybenzoate Impurity C
InChI Formula InChI=1S/C9H10O3/c1-2-12-9(11)7-3-5-8(10)6-4-7/h3-6,10H,2H2,1H3
SMILES CCOC(=O)c1ccc(O)cc1
IUPAC Name ethyl 4-hydroxybenzoate
Molecular Weight (g/mol) 166.17
Quantity 250 mg
Formula Weight 166.063 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.