missing translation for 'onlineSavingsMsg'
Learn More

Ethyl pentafluorobenzoate, 98%

Product Code. 11348485
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11348485

Brand: Thermo Scientific Alfa Aesar L10523.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 4522-93-4 | C9H5F5O2 | 240.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4522-93-4
Molecular Formula C9H5F5O2
Molecular Weight (g/mol) 240.13
MDL Number MFCD00039211
InChI Key DFUDMSIRGGTHGI-UHFFFAOYSA-N
Synonym ethyl pentafluorobenzoate, ethyl perfluorobenzoate, pentafluorobenzoic acid ethyl ester, 2,3,4,5,6-pentafluoro-benzoic acid ethyl ester, benzoic acid, pentafluoro-, ethyl ester, ethylpentafluorobenzoate, acmc-1ajj1, rarechem al bi 0017, perfluorobenzoic acid ethyl ester
PubChem CID 78283
IUPAC Name ethyl 2,3,4,5,6-pentafluorobenzoate
SMILES CCOC(=O)C1=C(F)C(F)=C(F)C(F)=C1F

Specifications

Density 1.431
Boiling Point 112°C to 114°C
Flash Point 80°C (176°F)
Refractive Index 1.428
Quantity 5 g
Beilstein 2054770
Formula Weight 240.13
Percent Purity 98%
Chemical Name or Material Ethyl pentafluorobenzoate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.