missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Salicylate, Mikromol™

Product Code. 17971696
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17971696 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17971696 Supplier Mikromol Supplier No. MM0045.16
 Restricted Item
Estimated Shipment: 23-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Ethyl Salicylate
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Salicylic acid ethyl ester
CAS 118-61-6
Analyte or Component Names Salicylic acid ethyl ester
Shipping Condition Room Temperature
Molecular Formula C9 H10 O3
Certifications/Compliance ISO 17025
Synonym 2-Hydroxybenzoic acid ethyl ester, Salicylic acid, ethyl ester (6CI,7CI,8CI), 2-Ethoxycarbonylphenol, 2-Hydroxybenzoic acid ethyl ester, Ethyl 2-hydroxybenzoate, Ethyl o-hydroxybenzoate, Ethyl salicylate, Mesotol, NSC 8209, Sal Ethyl, Salicylic ether, Salotan, o-(Ethoxycarbonyl)phenol, Methyl Salicylate Impurity F
InChI Formula InChI=1S/C9H10O3/c1-2-12-9(11)7-5-3-4-6-8(7)10/h3-6,10H,2H2,1H3
SMILES CCOC(=O)c1ccccc1O
IUPAC Name ethyl 2-hydroxybenzoate
Molecular Weight (g/mol) 166.17
Quantity 100 mg
Formula Weight 166.063 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.