missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Sulfate Sodium Salt, LoGiCal

Product Code. 30459525
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30459525 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30459525 Supplier LoGiCal Supplier No. LGCFOR0397.04

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.

TRUSTED_SUSTAINABILITY

Specifica

Chemical Name or Material Ethyl sulfate sodium salt (as ethyl sulfate)
CAS 546-74-7
Analyte or Component Names Ethyl sulfate sodium salt (as ethyl sulfate)
Shipping Condition Room Temperature
Molecular Formula C2 H5 O4 S . Na
Certifications/Compliance ISO 9001
Synonym Sulfuric acid, monoethyl ester, sodium salt (1:1), Ethyl sodium sulfate (6CI,7CI), Sulfuric acid, monoethyl ester, sodium salt (8CI,9CI), Sodium ethyl sulfate, Sodium sulfovinate, Ethyl Sulfate Sodium Salt
InChI Formula InChI=1S/C2H6O4S.Na/c1-2-6-7(3,4)5;/h2H2,1H3,(H,3,4,5);/q;+1/p-1
Solution Type Neat
SMILES [Na+].CCOS(=O)(=O)[O-]
IUPAC Name sodium;ethyl sulfate
Industry Type Clinical Testing
Molecular Weight (g/mol) 148.11
Quantity 10 mg
Formula Weight 147.9806 g/mol
Vedi altri risultati Mostra meno risultati
Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.