missing translation for 'onlineSavingsMsg'
Learn More

Ethyl vanillate, 97%

Product Code. p-7044314
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
This item is not returnable. View return policy

Product Code. 11397595

Brand: Thermo Scientific Alfa Aesar L05798.09

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 617-05-0 | C10H12O4 | 196.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 617-05-0
Molecular Formula C10H12O4
Molecular Weight (g/mol) 196.20
MDL Number MFCD00017269
InChI Key MWAYRGBWOVHDDZ-UHFFFAOYSA-N
Synonym ethyl vanillate, vanillic acid, ethyl ester, 4-hydroxy-3-methoxybenzoic acid ethyl ester, benzoic acid, 4-hydroxy-3-methoxy-, ethyl ester, unii-v38jk4z93o, 3-methoxy-4-hydroxybenzoic acid, ethyl ester, m-anisic acid, 4-hydroxy-, ethyl ester, vanilicacidethylester, ethyl 4-hydroxy-3-methoxy-benzoate, ethylvanillate
PubChem CID 12038
IUPAC Name ethyl 4-hydroxy-3-methoxybenzoate
SMILES CCOC(=O)C1=CC=C(O)C(OC)=C1

Specifications

Melting Point 42°C to 46°C
Boiling Point 293°C
Quantity 10 g
Beilstein 2100025
Formula Weight 196.2
Percent Purity 97%
Chemical Name or Material Ethyl vanillate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.