missing translation for 'onlineSavingsMsg'
Learn More

ethyle2-bromobenzoate, 97%, Maybridge™

Product Code. p-2704101
Change view
Click to view available options
Quantity:
10 g
50 g
100 g
Unit Size:
100g
10g
50g
Product Code. Quantity unitSize
10603075 10 g 10g
10032124 50 g 50g
10575855 100 g 100g
3 options
This item is not returnable. View return policy

Product Code. 10603075

Brand: Thermo Scientific Maybridge RDP00160EA

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 6091-64-1
Molecular Formula C9H9BrO2
Molecular Weight (g/mol) 229.073
InChI Key BIHHBTVQFPVSTE-UHFFFAOYSA-N
Synonym 2-bromobenzoic acid ethyl ester, benzoic acid, 2-bromo-, ethyl ester, ethyl-2-bromobenzoate, ethyl2-bromobenzoate, ethyl bromobenzoate, ethyl o-bromobenzoate, pubchem3937, brombenzoesaureathylester, acmc-209mn8, ksc493o4r
PubChem CID 80186
IUPAC Name ethyl 2-bromobenzoate
SMILES CCOC(=O)C1=CC=CC=C1Br

Specifications

CAS Min % 97.0
Density 1.44g/mL
Assay Percent Range 97%
Packaging Amber glass bottle
Linear Formula BrC6H4CO2C2H5
Quantity 10 g
Formula Weight 229.07
Percent Purity 97%
Chemical Name or Material ethyle2-bromobenzoate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.