missing translation for 'onlineSavingsMsg'
Learn More

ethyle3,5-dichlorobenzoate, 97%, Thermo Scientific™

Product Code. p-2704197
Click to view available options
Quantity:
1 g
10 g
25 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
25g
This item is not returnable. View return policy

Product Code. 10029521

Brand: Thermo Scientific Maybridge TL00759DA

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 91085-56-2
Molecular Formula C9H8Cl2O2
Molecular Weight (g/mol) 219.06
MDL Number MFCD00221467
InChI Key JRLNLVFPSMDPLU-UHFFFAOYSA-N
Synonym ethyl3,5-dichlorobenzoate, 3,5-dichlorobenzoic acid ethyl ester, rarechem al bi 0057, 5-ethoxycarbonyl-1,3-dichlorobenzene, benzoic acid, 3,5-dichloro-, ethyl ester, pubchem10323, ethyl 3,5-dichloro benzoate, ethyl 3,5-bis chloranyl benzoate, 3,5-dichlorobenzoic acid ethyl etster
PubChem CID 2736404
IUPAC Name ethyl 3,5-dichlorobenzoate
SMILES CCOC(=O)C1=CC(Cl)=CC(Cl)=C1

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Linear Formula Cl2C6H3C(O)OCH2CH3
Quantity 1 g
Formula Weight 219.07
Percent Purity 97%
Chemical Name or Material Ethyl 3, 5-dichlorobenzoate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.