missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethylsulfate sodium salt, Lipomed
Description
Ethylsulfate sodium salt
Specifications
Specifications
| Chemical Name or Material | Ethyl sulfate sodium salt (as ethyl sulfate) |
| CAS | 546-74-7 |
| Analyte or Component Names | Ethyl sulfate sodium salt (as ethyl sulfate) |
| Shipping Condition | Room Temperature |
| Molecular Formula | C2 H5 O4 S . Na |
| Synonym | Sulfuric acid, monoethyl ester, sodium salt (1:1), Ethyl sodium sulfate (6 CI,7 CI), Sulfuric acid, monoethyl ester, sodium salt (8 CI,9 CI), Sodium ethyl sulfate, Sodium sulfovinate, Ethyl Sulfate Sodium Salt |
| InChI Formula | InChI=1 S/C16H21NO2.ClH/c1-12(2)17-10-14(18)11-19-16-9-5-7-13-6-3-4-8-15(13)16;/h3-9,12,14,17-18 H,10-11H2,1-2H3;1 H |
| SMILES | [Na+].CCOS(=O)(=O)[O-] |
| IUPAC Name | sodium;ethyl sulfate |
| Industry Type | Pharmaceutical |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?