missing translation for 'onlineSavingsMsg'
Learn More

Etomidate 0.1 mg/ml in Methanol, LoGiCal

Product Code. 30457704
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30457704 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30457704 Supplier LoGiCal Supplier No. LGCAMP1264.0002
 Restricted Item
Estimated Shipment: 02-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Etomidate
CAS 33125-97-2
Analyte or Component Names Etomidate
Shipping Condition Room Temperature
Molecular Formula C14 H16 N2 O2
Certifications/Compliance ISO 17034
Synonym 1H-Imidazole-5-carboxylic acid, 1-[(1R)-1-phenylethyl]-, ethyl ester, 1H-Imidazole-5-carboxylic acid, 1-(1-phenylethyl)-, ethyl ester, (R)-, (+)-Etomidate, Amidate, Amidate (pharmaceutical), D-Etomidate, Etomidate, Hypnomidate, Lipuro, Propiscin, R 16659, Radenarcon, d-Etomidate, Imidazole-5-carboxylic acid, 1-(alpha-methylbenzyl)-, ethyl ester, (R)-(+)-
InChI Formula InChI=1S/C14H16N2O2/c1-3-18-14(17)13-9-15-10-16(13)11(2)12-7-5-4-6-8-12/h4-11H,3H2,1-2H3/t11-/m1/s1
Solution Type Single Solution
SMILES CCOC(=O)c1cncn1[C@H](C)c2ccccc2
IUPAC Name ethyl 3-[(1R)-1-phenylethyl]imidazole-4-carboxylate
Solvent or Matrix Methanol
Industry Type Clinical Testing
Molecular Weight (g/mol) 244.29
Quantity 1 mL
Formula Weight 244.1212 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.