missing translation for 'onlineSavingsMsg'
Learn More

Etomidate, Mikromol™

Product Code. 17950566
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17950566 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17950566 Supplier Mikromol Supplier No. MM1264.00
 Restricted Item
Estimated Shipment: 02-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Etomidate
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Etomidate
CAS 33125-97-2
Analyte or Component Names Etomidate
Shipping Condition Room Temperature
Molecular Formula C14 H16 N2 O2
Certifications/Compliance ISO 17034
Synonym 1H-Imidazole-5-carboxylic acid, 1-[(1R)-1-phenylethyl]-, ethyl ester, 1H-Imidazole-5-carboxylic acid, 1-(1-phenylethyl)-, ethyl ester, (R)-, (+)-Etomidate, Amidate, Amidate (pharmaceutical), D-Etomidate, Etomidate, Hypnomidate, Lipuro, Propiscin, R 16659, Radenarcon, d-Etomidate, Imidazole-5-carboxylic acid, 1-(alpha-methylbenzyl)-, ethyl ester, (R)-(+)-
InChI Formula InChI=1S/C14H16N2O2/c1-3-18-14(17)13-9-15-10-16(13)11(2)12-7-5-4-6-8-12/h4-11H,3H2,1-2H3/t11-/m1/s1
SMILES CCOC(=O)c1cncn1[C@H](C)c2ccccc2
IUPAC Name ethyl 3-[(1R)-1-phenylethyl]imidazole-4-carboxylate
Molecular Weight (g/mol) 244.29
Quantity 100 mg
Formula Weight 244.1212 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.