missing translation for 'onlineSavingsMsg'
Learn More

Etravirine, Thermo Scientific™

Product Code. 17392491 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17392491 100 mg 100mg
17302501 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17392491 Supplier Thermo Scientific Acros Supplier No. 468721000
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 269055-15-4
Molecular Formula C20H15BrN6O
Molecular Weight (g/mol) 435.29
MDL Number MFCD09837879
InChI Key PYGWGZALEOIKDF-UHFFFAOYSA-N
Synonym 4-((6-Amino-5-bromo-2-((4-cyanophenyl)amino)pyrimidin-4-yl)oxy)-3,5-dimethylbenzonitrile
IUPAC Name 4-({6-amino-5-bromo-2-[(4-cyanophenyl)amino]pyrimidin-4-yl}oxy)-3,5-dimethylbenzonitrile
SMILES CC1=CC(=CC(C)=C1OC1=NC(NC2=CC=C(C=C2)C#N)=NC(N)=C1Br)C#N

Specifications

Packaging Glass Bottle
Certificate of Analysis Available
Physical Form Powder
Color White to Yellow to Beige
Melting Point 262°C to 265°C
Solubility Insoluble in water
Quantity 100 mg
Formula Weight 435.28
Chemical Name or Material Etravirine
compliance-icons
Product Identifier
  • Etravirine
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
  • H400-Very toxic to aquatic life.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: Etravirine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.