missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Exonuclease 1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-68889
This item is not returnable.
View return policy
Description
Exonuclease 1 Polyclonal antibody specifically detects Exonuclease 1 in Human samples. It is validated for Immunocytochemistry/ Immunofluorescence
Specifications
| Exonuclease 1 | |
| Polyclonal | |
| Immunocytochemistry/ Immunofluorescence 0.25-2 μg/mL | |
| EXOI, exonuclease 1, Exonuclease I, HEX1rad2 nuclease family member, homolog of S. cerevisiae exonuclease 1, hExo1, hExoIEC 3.1 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: LLREGKVSEARECFTRSINITHAMAHKVIKAARSQGVDCLVAPYEADAQLAYLNKAGIVQAIITEDSDLLAFGCKKVILKMDQFGNGLEIDQARLGMCR | |
| 100 μg | |
| Apoptosis, Cancer, Core ESC Like Genes, DNA Repair, Editing and Processing Endonucleases, Stem Cell Markers, Tumor Suppressors | |
| 9156 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Immunofluorescence | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction