missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Exonuclease 1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
369.00 € - 559.00 €
Specifications
| Antigen | Exonuclease 1 |
|---|---|
| Dilution | Immunocytochemistry/ Immunofluorescence 0.25-2 μg/mL |
| Applications | Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18604177
|
Novus Biologicals
NBP2-68889-25ul |
25 μL |
369.00 €
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18615358
|
Novus Biologicals
NBP2-68889 |
100 μg |
559.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Exonuclease 1 Polyclonal antibody specifically detects Exonuclease 1 in Human samples. It is validated for Immunocytochemistry/ ImmunofluorescenceSpecifications
| Exonuclease 1 | |
| Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Apoptosis, Cancer, Core ESC Like Genes, DNA Repair, Editing and Processing Endonucleases, Stem Cell Markers, Tumor Suppressors | |
| PBS (pH 7.2) and 40% Glycerol | |
| 9156 | |
| IgG | |
| Protein A purified |
| Immunocytochemistry/ Immunofluorescence 0.25-2 μg/mL | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| EXOI, exonuclease 1, Exonuclease I, HEX1rad2 nuclease family member, homolog of S. cerevisiae exonuclease 1, hExo1, hExoIEC 3.1 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: LLREGKVSEARECFTRSINITHAMAHKVIKAARSQGVDCLVAPYEADAQLAYLNKAGIVQAIITEDSDLLAFGCKKVILKMDQFGNGLEIDQARLGMCR | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title