missing translation for 'onlineSavingsMsg'
Learn More

Exosome component 8 Antibody [Janelia Fluor« 669], Novus Biologicals Biologicals™

Product Code. 30495140 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30495140 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30495140 Supplier Novus Biologicals Supplier No. NBP338100JF669

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Exosome component 8 Polyclonal antibody specifically detects Exosome component 8 in Human samples. It is validated for ELISA,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen Exosome component 8
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Janelia Fluor 669
Formulation 50mM Sodium Borate
Gene Alias bA421P11.3, CBP-interacting protein 3, CIP3, EAP2, exosome complex exonuclease RRP43, exosome component 8Opa-interacting protein 2, OIP2Ribosomal RNA-processing protein 43, Opa interacting protein 2, p9RP11-421P11.3, RRP43OIP-2, Rrp43p
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-276 of human Exosome component 8 (NP_852480.1).,, Sequence:, MAAGFKTVEPLEYYRRFLKENCRPDGRELGEFRTTTVNIGSISTADGSALVKLGNTTVICGVKAEFAAPSTDAPDKGYVVPNVDLPPLCSSRFRSGPPGEEAQVASQFIADVIENSQIIQKEDLCISPGKLVWVLYCDLICLDYDGNILDACTFALLAALKNVQLPEVTINEETALAEVNLKKKSYLNIRTHPVATSFAVFDDTLLIVDPTGEEEHLATGTLTIVMDEEGKLCCLHKPGGSGLTGAKLQDCMSRAVTRHKEVKKLMDEVIKSMKPK
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline DNA replication Transcription Translation and Splicing
Primary or Secondary Primary
Gene ID (Entrez) 11340
Target Species Human
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.