missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DYKDDDDK Epitope Tag Antibody (L5), Novus Biologicals™
Rat Monoclonal Antibody has been used in 75 publications
Brand: Novus Biologicals NBP1-06712
This item is not returnable.
View return policy
Description
DYKDDDDK Epitope Tag Monoclonal specifically detects DYKDDDDK Epitope Tag in Epitope Tag samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Frozen).
Specifications
| DYKDDDDK Epitope Tag | |
| Monoclonal | |
| Unconjugated | |
| Tissue culture supernatant with 0.1% Sodium Azide | |
| Rat | |
| 1.01 kDa | |
| 0.5 mL | |
| ECS Epitope Tags (DDDDK), Epitope Tags | |
| Binds to same epitope as Sigma's Anti-FLAGM2 Antibody. FLAG is a registered trademark of Sigma-Aldrich Biotechnology LP and Sigma-Aldrich Co. | |
| Store at 4C. Do not freeze. | |
| IgG2a |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Frozen) | |
| L5 | |
| Western Blot 1:500-1:1000. Use reported in multiple pieces of scientific literature, Immunohistochemistry 1:50-1:100. Use reported in multiple pieces of scientific literature, Immunocytochemistry/Immunofluorescence 1:10-1:100. Use reported in multiple pieces of scientific literature, Immunoprecipitation reported in multiple pieces of scientific literature, Immunohistochemistry-Frozen reported in scientific literature (PMID 24454782) | |
| anti-flag L5, DDDDK epitope tag, DDK, DYKDDDDK tag, ECS epitope tag, ECS tag, Enterokinase Cleavage Site epitope tag, Enterokinase Cleavage Site tag, epitope tag flag, FLAG (FLAG is a trademark of Sigma Aldrich), Flag tag, Ty 1 Tag, xxxDDDDK epitope tag | |
| DYKDDDDK Epitope Tag Antibody (L5) was made to N-terminal DYKDDDDK-tagged extracellular domain of mouse Langerin. Binds to same epitope as SigmA′s Anti-FLAG® M2 Antibody. | |
| Tissue culture supernatant | |
| RUO | |
| Primary | |
| Guinea Pig | |
| Supernatant | |
| Epitope Tag |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction