missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DYKDDDDK Epitope Tag Antibody (L5), Novus Biologicals™
Rat Monoclonal Antibody has been used in 75 publications
264.00 € - 635.00 €
Specifications
| Antigen | DYKDDDDK Epitope Tag |
|---|---|
| Clone | L5 |
| Dilution | Western Blot 1:500-1:1000. Use reported in multiple pieces of scientific literature, Immunohistochemistry 1:50-1:100. Use reported in multiple pieces of scientific literature, Immunocytochemistry/Immunofluorescence 1:10-1:100. Use reported in multiple pieces of scientific literature, Immunoprecipitation reported in multiple pieces of scientific literature, Immunohistochemistry-Frozen reported in scientific literature (PMID 24454782) |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Frozen) |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18771354
|
Novus Biologicals
NBP1-06712SS |
0.125 mL |
264.00 €
125µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18788583
|
Novus Biologicals
NBP1-06712 |
0.5 mL |
635.00 €
0.50mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
DYKDDDDK Epitope Tag Monoclonal specifically detects DYKDDDDK Epitope Tag in Epitope Tag samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Frozen).Specifications
| DYKDDDDK Epitope Tag | |
| Western Blot 1:500-1:1000. Use reported in multiple pieces of scientific literature, Immunohistochemistry 1:50-1:100. Use reported in multiple pieces of scientific literature, Immunocytochemistry/Immunofluorescence 1:10-1:100. Use reported in multiple pieces of scientific literature, Immunoprecipitation reported in multiple pieces of scientific literature, Immunohistochemistry-Frozen reported in scientific literature (PMID 24454782) | |
| Monoclonal | |
| Supernatant | |
| RUO | |
| Guinea Pig | |
| anti-flag L5, DDDDK epitope tag, DDK, DYKDDDDK tag, ECS epitope tag, ECS tag, Enterokinase Cleavage Site epitope tag, Enterokinase Cleavage Site tag, epitope tag flag, FLAG (FLAG is a trademark of Sigma Aldrich), Flag tag, Ty 1 Tag, xxxDDDDK epitope tag | |
| IgG2a | |
| Tissue culture supernatant | |
| Binds to same epitope as Sigma's Anti-FLAGM2 Antibody. FLAG is a registered trademark of Sigma-Aldrich Biotechnology LP and Sigma-Aldrich Co. |
| L5 | |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Frozen) | |
| Unconjugated | |
| Rat | |
| ECS Epitope Tags (DDDDK), Epitope Tags | |
| Tissue culture supernatant with 0.1% Sodium Azide | |
| DYKDDDDK Epitope Tag Antibody (L5) was made to N-terminal DYKDDDDK-tagged extracellular domain of mouse Langerin. Binds to same epitope as SigmA′s Anti-FLAG® M2 Antibody. | |
| Primary | |
| Store at 4C. Do not freeze. | |
| 1.01 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title