missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
FAM117A Polyclonal specifically detects FAM117A in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
Specifications
| Antigen | FAM117A |
| Applications | Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1-4 ug/ml, Immunohistochemistry-Paraffin 1:500 - 1:1000 |
| Formulation | PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide |
| Gene Alias | C/EBP induced protein, C/EBP-induced protein, family with sequence similarity 117, member A |
| Gene Symbols | FAM117A |
| Host Species | Rabbit |
| Immunogen | This antibody was developed against Recombinant Protein corresponding to amino acids:KEISKLKQQLQRTKLSRSGKEKERGSPLLGDHAVRGALRASPPSFPSGSPVLRLSPCLHRSLEGLNQELEEVFVKEQGEEELLRILDIPDG |
| Visa mer |
For Research Use Only
Produkttitel
Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.
Hittar du en möjlighet till förbättring?