missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
FAM181B Polyclonal antibody specifically detects FAM181B in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
Specifications
| Antigen | FAM181B |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | mFluor Violet 450 SE |
| Dilution | Immunohistochemistry-Paraffin |
| Formulation | 50mM Sodium Borate |
| Gene Alias | Family With Sequence Similarity 181, Member B, Protein FAM181B |
| Host Species | Rabbit |
| Immunogen | Produced in rabbits immunized with E. coli-derived Human FAM181B fragment. |
| Purification Method | Antigen and protein A Affinity-purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?