missing translation for 'onlineSavingsMsg'
Learn More

Famotidine, Mikromol™

Product Code. 17902056
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17902056 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17902056 Supplier Mikromol Supplier No. MM0029.00
 Restricted Item
Estimated Shipment: 06-11-2026
Add to basket
Añadir a la cesta

missing translation for 'validMainframeMsg'

Este artículo no se puede devolver. Vea la política de devoluciones

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Famotidine
TRUSTED_SUSTAINABILITY

Especificaciones

Chemical Name or Material Famotidine
CAS 76824-35-6
Analyte or Component Names Famotidine
Shipping Condition Room Temperature
Molecular Formula C8 H15 N7 O2 S3
Certifications/Compliance ISO 17034
Synonym 3-[[[2-[(Aminoiminomethyl)amino]-4-thiazolyl]methyl]thio]-N-(aminosulfonyl)-propanimidamide, 3-[(2-Diaminomethyleneaminothiazol-4-yl)methylthio]-N-sulfamoylpropionamidine, Agufam, Amfamox, Antodine, Apo-Famotidine, Apogastine, Bestidine, Brolin, Cepal, Dibrit 40, Digervin, Dispromil, Dispronil, Durater, Facid, Fadin, Fadine, Fadyn, Fagastine, Famo, FamoABZ, Famocid, Famodar, Famodil, Famodin, Famodine, Famogard, Famogast, Famonerton, Famopsin, Famos, Famosan, Famostagine-D, Famotal, Famotep, Famotidine, Famotin, Famotine, Famowal, Famox, Famoxal, Fanosin, Fanox, Farmotex, Fenox, Fibonel, Fudone, Fuweidin, Ganor, Ganor mite, Gaster, Gastridin, Gastrion, Gastro, Gastrodomina, Gastropen, H 2 Bloc, Ifada, Kemofam, Lecedil, Logos, MK 208, Mosul, Motiax, Muclox, N-(Aminosulfonyl)-3-[[[2-[(diaminomethylene)amino]-4-thiazolyl]methyl]thio]propanimidamide, Nevofam, Nulcerin, Panalba, Pepcid, Pepcid AC, Pepcid PM, Pepcidin, Pepcidin Rapitab, Pepcidina, Pepcidine, Pepdif, Pepdine, Pepdul, Pepdul mite, Pepfamin, Peptan, Peptifam, Pepzan, Purifam, Quamatel, Quamtel, Restadin, Rogasti, Sedanium R, Supertidine, Tamin, Topcid, Ulcatif, Ulcedine, Ulceran, Ulcetrax, Ulcidine, Ulcofam, Ulfagel, Ulfam, Ulfamid, Ulfinol, Ulped, Ulped AR, Weimok, YM 11170
InChI Formula InChI=1S/C8H15N7O2S3/c9-6(15-20(12,16)17)1-2-18-3-5-4-19-8(13-5)14-7(10)11/h4H,1-3H2,(H2,9,15)(H2,12,16,17)(H4,10,11,13,14)
SMILES NC(=Nc1nc(CSCCC(=NS(=O)(=O)N)N)cs1)N
IUPAC Name 3-[[2-(diaminomethylideneamino)-1,3-thiazol-4-yl]methylsulfanyl]-N'-sulfamoylpropanimidamide
Molecular Weight (g/mol) 337.45
Quantity 250 mg
Formula Weight 337.0449 g/mol
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.