missing translation for 'onlineSavingsMsg'
Learn More

Fbx32 Antibody [PerCP], Novus Biologicals Biologicals™

Product Code. 30490839 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30490839 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30490839 Supplier Novus Biologicals Supplier No. NBP338033PCP

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Fbx32 Polyclonal antibody specifically detects Fbx32 in Mouse samples. It is validated for ELISA,Western Blot,Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen Fbx32
Applications ELISA, Western Blot, Immunocytochemistry/Immunofluorescence
Classification Polyclonal
Conjugate PerCP
Formulation PBS
Gene Alias atrogin 1, atrogin-1, ATROGIN1, F-box only protein 32, F-box protein 32, Fbx32, FLJ32424, MAFBX, MAFbxMGC33610, Muscle atrophy F-box protein
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 206-355 of human Fbx32 (NP_478136.1).,, Sequence:, WQQQLNNIQITRPAFKGLTFTDLPLCLQLNIMQRLSDGRDLVSLGQAAPDLHVLSEDRLLWKKLCQYHFSERQIRKRLILSDKGQLDWKKMYFKLVRCYPRKEQYGDTLQLCKHCHILSWKGTDHPCTANNPESCSVSLSPQDFINLFKF
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cardiovascular Biology, Cell Biology, Stem Cells
Primary or Secondary Primary
Gene ID (Entrez) 114907
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.